EMAIL THIS PAGE TO A FRIEND
Dermatology (Basel, Switzerland)
Drug rash with eosinophilia and systemic symptoms caused by topical application of mercury.
PMID 11146363
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M7011
Mercury dibromofluorescein disodium salt, practical grade, crystalline
C20H8Br2HgNa2O6