EMAIL THIS PAGE TO A FRIEND
Nature cell biology
A novel mitochondrial septin-like protein, ARTS, mediates apoptosis dependent on its P-loop motif.
PMID 11146656
Abstract
Here we describe a protein product of the human septin H5/PNUTL2/CDCrel2b gene, which we call ARTS (for apoptosis-related protein in the TGF-beta signalling pathway). ARTS is expressed in many cells and acts to enhance cell death induced by TGF-beta or, to a lesser extent, by other apoptotic agents. Unlike related septin gene products, ARTS is localized to mitochondria and translocates to the nucleus when apoptosis occurs. Mutation of the P-loop of ARTS abrogates its competence to activate caspase 3 and to induce apoptosis. Taken together, these observations expand the functional attributes of septins previously described as having roles in cytokinesis and cellular morphogenesis.
Related Materials