EMAIL THIS PAGE TO A FRIEND
FEBS letters
Endothelin-1[1-31], acting as an ETA-receptor selective agonist, stimulates proliferation of cultured rat zona glomerulosa cells.
PMID 11150508
Abstract
Endothelin-1 (ET-1)[1-31] is a novel hypertensive peptide that mimics many of the vascular effects of the classic 21 amino acid peptide ET-1[1-21]. However, at variance with ET-1[1-21] that enhances aldosterone secretion from cultured rat zona glomerulosa (ZG) cells by acting via ETB receptors, ET-1[1-31] did not elicit such effect. Both ET-1[1-21] and ET-1[1-31] raised the proliferation rate of cultured ZG cells, the maximal effective concentration being 10(-8) M. This effect was blocked by the ETA-receptor antagonist BQ-123 and unaffected by the ETB-receptor antagonist BQ-788. Quantitative autoradiography showed that ET-1[1-21] displaced both [(125)I]PD-151242 binding to ETA receptors and [(125)I]BQ-3020 binding to ETB receptors in both rat ZG and adrenal medulla, while ET-1[1-31] displaced only [(125)I]BQ-3020 binding. The tyrosine kinase (TK) inhibitor tyrphostin-23 and the p42/p44 mitogen-activated protein kinase (MAPK) inhibitor PD-98059 abolished the proliferogenic effect of ET-1[1-31], while the protein kinase-C (PKC) inhibitor calphostin-C significantly reduced it. ET-1[1-31] (10(-8) M) stimulated TK and MAPK activity of dispersed ZG cells, an effect that was blocked by BQ-123. The stimulatory action of ET-1[1-31] on TK activity was annulled by tyrphostin-23, while that on MAPK activity was reduced by calphostin-C and abolished by either tyrphostin-23 and PD-98059. These data suggest that ET-1[1-31] is a selective agonist of the ETA-receptor subtype, and enhances proliferation of cultured rat ZG cells through the PKC- and TK-dependent activation of p42/p44 MAPK cascade.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SCP0138 BQ-3020
C96H140N20O25S1
P208
PD 151,242, ≥97%, powder
C34H45N5O6