EMAIL THIS PAGE TO A FRIEND
Mechanisms of ageing and development
On the anti-aging activities of aminoguanidine and N-t-butylhydroxylamine.
PMID 11166356
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
194751
N-tert-Butylhydroxylamine hydrochloride, ≥98%
C4H12ClNO