EMAIL THIS PAGE TO A FRIEND
Chemical research in toxicology
Studies of the chemical selectivity of hapten, reactivity, and skin sensitization potency. 2. nmr studies of the covalent binding of the (13)c-labeled skin sensitizers 2-[13C]- and 3-[13C]hex-1-ene- and 3-[13C]hexane-1,3-sultones to human serum albumin.
PMID 11170515
Abstract
3-[(13)C]- and 2-[(13)C]hex-1-ene-1,3-sultones (1a and 1b, respectively) and 3-[(13)C]hex-1-ene-1,3-sultone 2a were incubated with human serum albumin in phosphate buffer at pH 8.1. In both cases, the main reaction was a hydrolysis via an S(N) reaction at position 3, but several adducts were also formed. Hex-1-ene-1,3-sultone, which is a strong skin sensitizer, appears also to be a strongly oxophilic molecule reacting mainly at position 3 through an S(N) reaction to give adducts on tyrosines. This sultone was also able to react with a single lysine residue, also via an initial S(N) reaction at position 3, followed by an intramolecular Michael addition at position 2 to form a mixture of aziridinium intermediates which were subsequently hydrolyzed to give an amino alcohol derivative as the final product. The same reaction carried out on acetylated human serum albumin seems to indicate that the target lysine could be Lys199, which is known to be easily acetylated. Hexane-1,3-sultone, which is a weak sensitizer, appears to be an even more oxophilic molecule, making adducts on tyrosines through an S(N) reaction at position 3. No reaction was observed on Lys199. The difference in skin sensitization potential seems therefore to be more related to the selective ability of modifying lysine residues than to the more general ability to modify tyrosine residues.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
316539
1,8-Naphthosultone, 98%
C10H6O3S