EMAIL THIS PAGE TO A FRIEND
The Journal of pediatrics
In vivo evidence of brain galactitol accumulation in an infant with galactosemia and encephalopathy.
PMID 11174626
Abstract
In a newborn infant with galactose-1-phosphate uridyltransferase deficiency and encephalopathy, brain magnetic resonance imaging revealed cytotoxic edema in white matter. Using in vivo proton magnetic resonance spectroscopy, we detected approximately 8 mmol galactitol per kilogram of brain tissue, an amount potentially relevant to the pathogenesis of brain edema.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D0256
Dulcitol, ≥99%
C6H14O6