EMAIL THIS PAGE TO A FRIEND
Bulletin of environmental contamination and toxicology
Effects of laundering upon the removal of atrazine and metolachlor from cotton, cotton/polyester, and polyester fabrics treated with fluorochemical finishes.
PMID 11178646