EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
2,2-Dimethyl-4,5-diaryl-3(2H)furanone derivatives as selective cyclo-oxygenase-2 inhibitors.
PMID 11206450
Abstract
A series of 2,2-dimethyl-5-[4-(methylsulfonyl)phenyl]-4-phenyl-3(2H)furanones was prepared and evaluated for their ability to inhibit cyclo-oxygenase-2 (COX-2).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
222771
4-(Methylthio)benzaldehyde, 95%
C8H8OS