EMAIL THIS PAGE TO A FRIEND
Pediatrics international : official journal of the Japan Pediatric Society
Precocious puberty in a girl with congenital hypothyroidism receiving continuous L-thyroxine-replacement therapy.
PMID 11208009