EMAIL THIS PAGE TO A FRIEND
Circulation
An HMG-CoA reductase inhibitor, cerivastatin, suppresses growth of macrophages expressing matrix metalloproteinases and tissue factor in vivo and in vitro.
PMID 11208689
Abstract
Unstable atherosclerotic plaques that cause acute coronary events usually contain abundant macrophages expressing matrix metalloproteinases (MMPs) and tissue factor (TF), molecules that probably contribute to plaque rupture and subsequent thrombus formation. Lipid lowering with HMG-CoA reductase inhibitors reduces acute coronary events. To test whether lipid lowering with an HMG-CoA reductase inhibitor retards macrophage accumulation in rabbit atheroma, we administered cerivastatin to immature Watanabe heritable hyperlipidemic rabbits (cerivastatin group, n=10, cerivastatin 0.6 mg x kg(-1) x d(-1); control group, n=9, saline 0.6 mL x kg(-1) x d(-1)) for 32 weeks and measured macrophage accumulation and expression of MMPs and TF. Serum cholesterol levels after 32 weeks were 809+/-40 mg/dL (control group) and 481+/-24 mg/dL (treated group). Cerivastatin diminished accumulation of macrophages in aortic atheroma. Macrophage expression of MMP-1, MMP-3, MMP-9, and TF also decreased with cerivastatin treatment. Cerivastatin reduced the number of macrophages expressing histone mRNA (a sensitive marker of cell proliferation) detected by in situ hybridization but did not alter macrophages bearing a marker of death (TUNEL staining). Cerivastatin treatment (>or=0.01 micromol/L) also reduced growth, proteolytic activity due to MMP-9, and TF expression in cultured human monocyte/macrophages. These results suggest that lipid lowering with HMG-CoA reductase inhibitors alters plaque biology by reducing proliferation and activation of macrophages, prominent sources of molecules responsible for plaque instability and thrombogenicity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0005
Cerivastatin sodium salt hydrate, ≥98% (HPLC)
C26H33FNNaO5 · xH2O