EMAIL THIS PAGE TO A FRIEND
Journal of neurochemistry
Peroxynitrite affects Ca2+ influx through voltage-dependent calcium channels.
PMID 11208897
Abstract
The effect of peroxynitrite (OONO-) on voltage-dependent Ca2+ channels (VDCCs) was examined by measuring [45Ca2+] influx into mouse cerebral cortical neurones. OONO- time- and dose-dependently increased [45Ca2+] influx and this increase was abolished by manganese (III) tetrakis (4-benzoic acid) porphyrin, a scavenger for OONO-. Inhibition of cyclic GMP (cGMP) formation did not alter the OONO(-)-induced [45Ca2+] influx. OONO-, as well as 30 mm KCl, significantly increased fluorescence intensity of cell-associated bis-(1,3-dibutylbarbituric acid) trimethine oxonol (bis-oxonol). Tetrodotoxin and membrane stabilizers such as lidocaine dose-dependently suppressed OONO(-)-induced [45Ca2+] influx. Although each of 1 microM nifedipine and 1 microM omega-agatoxin VIA (omega-ATX) significantly inhibited the OONO(-)-induced [45Ca2+] influx and the concomitant presence of these agents completely abolished the influx, 1 microM omega-conotoxin GVIA (omega-CTX) showed no effect on the influx. On the other hand, OONO- itself reduced 30 mM KCl-induced [45Ca2+] influx to the level of [45Ca2+] influx induced by OONO- alone, and the magnitude of this reduction was as same as that of KCl-induced [45Ca2+] influx by omega-CTX. These results indicate that OONO- increases [45Ca2+] influx into the neurones through opening P/Q- and L-type VDCCs subsequent to depolarization, and inhibits the influx through N-type VDCCs.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
44977
DISBAC2(3), Voltage Sensitive Probe, ≥98% (HPLC)
C19H24N4O4S2