EMAIL THIS PAGE TO A FRIEND
Medizinische Klinik (Munich, Germany : 1983)
[Cerebral calcinosis].
PMID 11210491
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D9257
Dihydrotachysterol, analytical standard
C28H46O