EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Nipecotic and iso-nipecotic amides as potent and selective somatostatin subtype-2 receptor agonists.
PMID 11212124
Abstract
N-Substituted nipecotic and iso-nipecotic amides of beta-methylTrpLys tert-butyl ester were found to be novel, selective and potent agonists of the somatostatin subtype-2 receptor in vitro. For example iso-nipecotic amide 8a showed high hsst2 binding affinity (Ki = 0.5 nM) and good selectivity (h5/h2 = 832).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N8105
Nipecotamide, 95%
C6H12N2O