EMAIL THIS PAGE TO A FRIEND
Journal of investigative medicine : the official publication of the American Federation for Clinical Research
Beneficial intervention of experimental colitis by passive cigarette smoking through the modulation of cytokines in rats.
PMID 11217144
Abstract
Epidemiologic observations have indicated that cigarette smoking decreases the risk of ulcerative colitis, but the modes of action remain anonymous. The present study aimed to investigate the beneficial effects of passive cigarette smoking using an animal colitis model. We hypothesized that the underlying mechanisms may involve immunoregulation of cytokines. Experimental colitis was induced in rats by enema administration of 2,4-dinitrobenzene sulfonic acid (DNBS). Passive cigarette smoking by rats was performed for 1 hour once daily, from 3 days before DNBS enema until they were sacrificed on day 8. Other groups of DNBS-treated rats received therapeutic treatment of cyclosporin A or pentoxifylline, a tumor necrosis factor (TNF)-alpha inhibitor. Macroscopic and histologic damage were graded, and the colonic levels of different cytokines and the levels/activities of parameters related to neutrophil activation were also measured. DNBS-induced colonic damage was improved in passive-cigarette-smoking rats. This was accompanied by attenuation of the elevated colonic myeloperoxidase and inducible nitric oxide synthase activities and leukotriene B4 level. Likewise, the augmentation in colonic levels of TNF-alpha, interleukin (IL)-1 beta, and IL-6 in colitis rats was also alleviated by passive cigarette smoking. In contrast, the deprivation of colonic IL-10 during colitis was preserved in cigarette-smoking rats. These effects were similarly accomplished by pentoxifylline and, to some degree, by cyclosporin A. The results support the idea that the beneficial effects of passive cigarette smoking in experimental colitis involved immunoregulation of cytokines in colonic tissues.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
259934
Sodium 2,4-dinitrobenzenesulfonate, 98%
C6H3N2NaO7S