EMAIL THIS PAGE TO A FRIEND
Carbohydrate research
Acid-catalyzed dehydrative cyclization of 4-(D-galacto -pentitol-1-yl)-2-phenyl-2H-1,2,3-triazole. synthesis and anomeric configuration of D-lyxo-C-nucleoside analogs.
PMID 11217962
Abstract
Dehydration of 4-(D-galacto-pentitol-1-yl)-2-phenyl-2H-1,2,3-triazole with 20% methanolic sulfuric acid afforded the anomeric pairs of nucleosides, 4-(alpha-D-lyxopyranosyl)-2-phenyl-2H-1,2,3-triazole (major component) and its beta-anomer, as well as 4-(alpha-D-lyxofuranosyl)-2H-1,2,3-triazole and its beta-anomer. The four anomeric C-nucleosides were separated by chromatography, and their structure and anomeric configuration were determined by periodate oxidation, acylation, and NMR spectroscopy as well as mass spectrometry. The anomeric assignment from optical rotation was not in agreement with final structure assignment and represented a violation of the Hudson isorotation rules. NOE studies and X-ray diffraction measurements confirmed the anomeric configuration.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
318183
Methyl sulfate sodium salt
CH3NaO4S