EMAIL THIS PAGE TO A FRIEND
Analytical biochemistry
Gas chromatographic determination of polyols and aldoses in human urine as polyacetates and aldononitrile polyacetates.
PMID 1122027
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
91204
Acetic anhydride, for GC derivatization, ≥99.0%
C4H6O3