EMAIL THIS PAGE TO A FRIEND
Microbiology and immunology
Effect of sulbactam on anti-pseudomonal activity of beta-lactam antibiotics in cells producing various levels of the MexAB-OprM efflux pump and beta-lactamase.
PMID 11220688
Abstract
The beta-lactamase inhibitor, sulbactam, was tested for beta-lactamase inhibitory activity in Pseudomonas aeruginosa cells producing various levels of both the MexAB-OprM efflux pump and beta-lactamase. We found that sulbactam lowered the MICs of cefoperazone and piperacillin by inhibiting the beta-lactamase 8-fold in the cell producing a constitutively high level of AmpC-type beta-lactamase and a wild-type level of MexAB-OprM pump compared with that without sulbactam. The MICs of cefoperazone and piperacillin in the cell producing a constitutively high level of both the efflux pump and beta-lactamase under the presence of sulbactam were 8 and 4 times, respectively, lower than that without sulbactam. The MICs of sulbactam in the cell producing a constitutively high and a wild-type level of the efflux pump were 16- and 8-fold higher, respectively, than that in the mutant lacking the efflux pump. We concluded that sulbactam exerts potent beta-lactamase inhibitory activity in the cell producing a high level of efflux pump, in spite of the fact that sulbactam serves as a substrate of the MexAB-OprM pump. Increasing amounts of sulbactam over the weight of beta-lactams further strengthen the effect of beta-lactam antibiotics.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C8145
Cefsulodin sodium salt hydrate, third-generation cephalosporin antibiotic
C22H19N4NaO8S2 · xH2O