EMAIL THIS PAGE TO A FRIEND
Diabetes, obesity & metabolism
Metabolic effects of moxonidine and other centrally acting antihypertensives.
PMID 11225647
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
Y0000226 Moxonidine, European Pharmacopoeia (EP) Reference Standard
C9H12ClN5O