EMAIL THIS PAGE TO A FRIEND
Clinica chimica acta; international journal of clinical chemistry
Determination of phosphate in serum and urine by a single step malachite-green method.
PMID 1122646
Abstract
The highly sensitive malachite-green method for the determination of phosphate requires a protective colloid in order to avoid precipitation of the formed dye salt. A polyvinylalcohol proved to be suitable and permit the determination of phosphate in urine and in serum without prior precipitation of the proteins. The present method is suitable for a one-step procedure which allows the analysis of several hundred samples per man-hour and is applicable to microquantities of serum or urine.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
32745
Malachite Green oxalate salt, for microscopy, crystalline, S. No.: 754
C23H25N2 · C2HO4 · 0.5C2H2O4