EMAIL THIS PAGE TO A FRIEND
The EMBO journal
A novel mRNA-decapping activity in HeLa cytoplasmic extracts is regulated by AU-rich elements.
PMID 11230136
Abstract
While decapping plays a major role in mRNA turnover in yeast, biochemical evidence for a similar activity in mammalian cells has been elusive. We have now identified a decapping activity in HeLa cytoplasmic extracts that releases (7me)GDP from capped transcripts. Decapping is activated in extracts by the addition of (7me)GpppG, which specifically sequesters cap-binding proteins such as eIF4E and the deadenylase DAN/PARN. Similar to in vivo observations, the presence of a poly(A) tail represses decapping of RNAs in vitro in a poly(A)-binding protein-dependent fashion. AU-rich elements (AREs), which act as regulators of mRNA stability in vivo, are potent stimulators of decapping in vitro. The stimulation of decapping by AREs requires sequence-specific ARE-binding proteins. These data suggest that cap recognition and decapping play key roles in mediating mRNA turnover in mammalian cells.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M5883
7-Methylguanosine 5′-diphosphate sodium salt, ≥92.5%
C11H17N5O11P2 · xNa+