EMAIL THIS PAGE TO A FRIEND
The Journal of clinical endocrinology and metabolism
The melanocortin melanocyte-stimulating hormone/adrenocorticotropin(4-10) decreases body fat in humans.
PMID 11238499
Abstract
The control of body fat is a prominent factor in human health. Animal studies have indicated a homeostatic central nervous system regulation of body fat with particular involvement of the melanocortin receptor pathway. This study provides evidence for a similar role for melanocortins in the long-term control of fat stores in humans. Thirty-six normal weight humans were assigned to one of three experimental groups. After a 4-week baseline, one group was treated with MSH/ACTH(4-10) (MSH/ACTH(4-10)) representing the core sequence of all melanocortins. Another group received desacetyl-alphaMSH, a selective agonist of the brain melanocortin-4 receptor, which shares the 4-10 sequence with MSH/ACTH(4-10). The third group received placebo. Treatments were given intranasally twice daily for 6 weeks, at equimolar doses (MSH/ACTH(4-10), 0.5 mg; desacetyl-alphaMSH, 0.84 mg). Body weight, body composition, and plasma hormone concentrations were measured before and after treatment. MSH/ACTH(4-10) reduced body fat, on the average, by 1.68 kg (P < 0.05) and body weight by 0.79 kg (P < 0.001). Concurrently, plasma leptin levels were decreased by 24% (P < 0.02), and insulin levels were decreased by 20% (P< 0.05) after MSH/ACTH(4-10). Changes after desacetyl-alphaMSH remained nonsignificant. The finding of reduced body adiposity after MSH/ACTH(4-10) confirms and extends to the human the findings of animal models indicating an essential role of the hypothalamic melanocortin system in body weight control.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A0401
Adrenocorticotropic Hormone Fragment 4-10 human, rat, ≥97% (HPLC)
C44H59N13O10S