EMAIL THIS PAGE TO A FRIEND
Journal of environmental monitoring : JEM
Spectrophotometric determination of ferbam (iron(III) dimethyldithiocarbamate) in commercial sample and wheat grains using 4,7-diphenyl-1,10-phenanthroline after extraction into mesityl oxide.
PMID 11253035
Abstract
A procedure has been developed for the determination of iron(III) dimethyldithiocarbamate by converting it into iron(II)-bathophenanthroline complex, which is then extracted into mesityl oxide in the presence of potassium perchlorate. The absorbance is then measured at 534 nm against a reagent blank. Beer's law is obeyed over the concentration range 0.5-20 microg ml(-1) in the final solution. The method is sensitive, highly selective and can be used for the determination of ferbam in a commercial sample, and in mixtures with various dithiocarbamates (ziram, zineb, maneb, etc.) and in wheat grains.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
W336807
4-Methyl-3-penten-2-one, 90%
C6H10O
281832
Mesityl oxide, technical grade, 90%
C6H10O