EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. B, Biomedical sciences and applications
Determination of aloenin, barbaloin and isobarbaloin in aloe species by micellar electrokinetic chromatography.
PMID 11254203
Abstract
Aloenin, barbaloin and isobarbaloin in JP Aloe, Aloe barbadensis (Aloe vera) and Aloe arborescens Miller var. natalensis Berger (Aloe arborescens Miller) were determined by micellar electrokinetic chromatography (MEKC) with 50 mM sodium dodecyl sulfate. Aloenin, barbaloin and isobarbaloin were well separated by MEKC and as little as 5.5 pg/11 nl of the three compounds could be detected. The determination took around 14 min.