EMAIL THIS PAGE TO A FRIEND
Organic letters
An approach to the skeleton of the securinega alkaloids. The total synthesis of (+/-)-securinine.
PMID 11259041
Abstract
[structure: see text]. The concise total synthesis of securinine in nine steps from readily available starting materials is described. Key steps of the synthesis include an addition of a silyloxyfuran to an in situ generated iminium ion and a novel ring closing metathesis reaction.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0055
Securinine, ≥98% (HPLC)
C13H15NO2