EMAIL THIS PAGE TO A FRIEND
Revista de investigacion clinica; organo del Hospital de Enfermedades de la Nutricion
[Advertising in the Revista de Investigación Clínica].
PMID 11261439
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G7012
Guaiacol glyceryl ether carbamate, analytical standard
C11H15NO5