EMAIL THIS PAGE TO A FRIEND
Journal of agricultural and food chemistry
Development of a gas chromatographic method for fungicide cymoxanil analysis in dried hops.
PMID 11261994
Abstract
An analytical method for detecting cymoxanil [2-cyano-N-[(ethylamino)carbonyl]-2-(methoxyimino)acetamide] residues in dried hops was developed utilizing liquid-liquid partitioning, automated gel permeation chromatography (GPC), solid phase extraction (SPE) cleanup, and gas chromatography (GC). Method validation recoveries from dried hops were 96 +/- 12, 108 +/- 11, and 136 +/- 8% over three levels of fortification (0.05, 0.5, and 1.0 ppm, respectively). The hop samples from three field sites, which were treated with cymoxanil, had residue levels ranging from 0.146 to 0.646 ppm. The detection limit and the quantitation limit of the method developed in the present study were 0.022 and 0.050 ppm, respectively.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
34326
Cymoxanil, PESTANAL®, analytical standard
C7H10N4O3