EMAIL THIS PAGE TO A FRIEND
Journal of agricultural and food chemistry
Residue levels of chlorpropham in individual tubers and composite samples of postharvest-treated potatoes.
PMID 11262017
Abstract
Chlorpropham, a herbicide and sprout suppressant, is used on stored potatoes to prolong the storage period without deterioration of produce quality. Data for residue concentrations on an individual tuber basis are required by WHO for the estimation of the variability factor. In this study, the levels of chlorpropham in individual tubers and in composite samples were determined. The distribution of chlorpropham between the peel and the tuber flesh was examined, and the fate during the cooking process (washing, boiling, frying) was studied. The concentrations in individual tubers ranged from 1.8 to 7.6 mg/kg 10 days postapplication (mean 3.8 mg/kg, RSD 39%), from 0.7 to 4.0 mg/kg 28 days postapplication (mean 2.9 mg/kg, RSD 28%), and from 0.8 to 3.8 mg/kg 65 days postapplication (mean 2.2 mg/kg, RSD 48%). The calculated residues in composite samples 10 days postapplication ranged from 4.3 to 6.1 mg/kg (mean 4.9 mg/kg, RSD 20%). Those in samples taken 28 days postapplication ranged from 3.1 to 4.2 mg/kg (mean 3.8 mg/kg, RSD 15%). The concentrations determined in composite samples of whole tubers 65 days postapplication ranged between 2.6 and 3.2 (mean 2.9 mg/kg, RSD 11%). Peeling removed 91-98% of the total residue; washing reduced residues by 33-47%. Detectable residues were found in boiled potatoes and the boiling water, and in french fries and the frying oil. Monitoring data on commercial prefried frozen french fries are reported.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
45393
Chlorpropham, PESTANAL®, analytical standard
C10H12ClNO2