EMAIL THIS PAGE TO A FRIEND
Biomedical chromatography : BMC
Stereoselective determination of p-hydroxyphenyl-phenylhydantoin enantiomers in rat liver microsomal incubates by reversed-phase high-performance liquid chromatography using beta-cyclodextrin as chiral mobile phase additives.
PMID 11268054
Abstract
An analytical method was developed for determination of p-hydroxyphenylphenylhydantoin enantiomers in rat liver microsome by using reversed-phase high-performance liquid chromatography. A 50 mm C(8) column was used as the analytical column. The mobile phase was made up of 8.8 mmol/L beta-cyclodextrin, 0.25 mol/L urea and 0.05 mol/L ammonium acetate in water. The assay was linear from 2.05 to 410.0 micromol/L for each enantiomer. The limits of detection and of quantitation for the method were 0.90 and 2.05 micromol/L for each enantiomer, respectively. The analytical method afforded average recoveries of 93.59 +/- 2.75% and 94.72 +/- 1.78% for S- and R-p-hydroxyphenylphenylhydantoin, respectively. The method allowed study of the in vitro glucuronidation of p-hydroxyphenylphenylhydantoin in rat liver microsomal incubates. The stereoselectivity of p-hydroxyphenylphenylhydantoin phase II metabolism was observed.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
161543
5-(4-Hydroxyphenyl)-5-phenylhydantoin, 98%
C15H12N2O3