EMAIL THIS PAGE TO A FRIEND
American journal of kidney diseases : the official journal of the National Kidney Foundation
Should we still use iron dextran in hemodialysis patients?
PMID 11273888
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D8517 Iron–Dextran, solution