EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
Highly regioselective Vilsmeier-Haack acylation of hexahydropyrroloindolizine.
PMID 11281803
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H3648
(±)-3-Hydroxydecanoic acid, ≥98%
C10H20O3