Description
Molecular Formula
Add to Cart
P8810
Phytic acid sodium salt hydrate, from rice
C6H18O24P6 · xNa+ · yH2O
68388
Phytic acid sodium salt hydrate, ≥90% phosphorus (P) basis (dry basis)
C6H18O24P6 · xNa+ · yH2O