EMAIL THIS PAGE TO A FRIEND
Neuroscience Letters
Glycogen synthase kinase-3beta immunoreactivity is reduced in the prefrontal cortex in schizophrenia.
PMID 11290401
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
AB5972 Anti-Dishevelled-2 Antibody, clone, Chemicon®, from rabbit