EMAIL THIS PAGE TO A FRIEND
Acta microbiologica Polonica
Demethylation of [14C]-labelled veratric acid and oxidation of methanol and formaldehyde by the white-rot fungus Phlebia radiata.
PMID 11293653
Abstract
Veratric acids 14C-labelled in carboxyl group, 3-OCH3, 4-OCH3, or aromatic ring together with unlabelled veratric acid were supplemented in the cultures of the white-rot fungus Phlebia radiata. The effect of various carbon sources on the release of 14CO2 was studied. Veratric acid was readily decarboxylated, maximally already on day 1 from the addition of [14COOH]-veratric acid. High amounts (4%) of glucose slightly repressed the decarboxylation. In medium supplemented with cellulose the methoxyl group in position 4 was much more readily mineralized to CO2 than the group in position 3. The maximum evolution was achieved on day 5, two days from the addition. Cellulose did not repress methanol oxidation but repression of methanol oxidation by glucose was detected in media supplemented with [O14CH3]-veratric acids and 14CH3OH. However, glucose did not repress oxidation of H14CHO. The apparent uptake of 14C by fungal mycelium, especially from methoxyl groups, but also from the aromatic ring, may partially be due to the strong slime formation observed in cellobiose medium. Also in cellobiose medium apparent uptake of 14C from 14C-labelled methoxyl groups was observed.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D131806
3,4-Dimethoxybenzoic acid, ≥99%
C9H10O4