EMAIL THIS PAGE TO A FRIEND
Journal of the Indian Medical Association
Use of anafortan intravenous injection for treatment of colicky pain.
PMID 11294338
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C4665
Camylofine dihydrochloride, analytical standard, for drug analysis
C19H32N2O2 · 2HCl