Glycoconjugate journal
Evaluation of synthetic schemes to prepare immunogenic conjugates of Vibrio cholerae O139 capsular polysaccharide with chicken serum albumin.
PMID 11294508
Vibrio cholerae serotype O139 is a new etiologic agent of epidemic cholera. There is no vaccine available against cholera caused by this serotype. V. cholerae O139 is an encapsulated bacterium, and its polysaccharide capsule is an essential virulent factor and likely protective antigen. This study evaluated several synthetic schemes for preparation of conjugates of V. cholerae O139 capsular polysaccharide (CPS) with chicken serum albumin as the carrier protein (CSA) using 1-ethyl-3(3-dimethylaminopropyl)carbodiimide (EDC) or 1-cyano-4-dimethylaminopyridinium tetrafluoroborate (CDAP) as activating agents. Four conjugates described here as representative of many experiments were synthesized in 2 steps: 1) preparation of adipic acid hydrazide derivative of CPS (CPS(AH)) or of CSA (CSA(AH)), and 2) binding of CPS(AH) to CSA or of CPS to CSA(AH). Although all conjugates induced CPS antibodies, the conjugate prepared by EDC-mediated binding of CPS and CSA(AH) (EDC:CPS-CSA(AH)) was statistically significantly less immunogenic than the other three conjugates. Representative sera from mice injected with these three conjugates contained antibodies that mediated the lysis of V. cholerae O139 inoculum. Evaluation of the different synthetic schemes and reaction conditions in relation to the immunogenicity of the resultant conjugates provided the basis for the preparation of a V. cholerae O139 conjugate vaccine with a medically useful carrier protein such as diphtheria toxin mutant.
Related Materials