EMAIL THIS PAGE TO A FRIEND
Biochemistry
Regulation of type II phosphatidylinositol phosphate kinase by tyrosine phosphorylation in bovine rod outer segments.
PMID 11294622
Abstract
Type II phosphatidylinositol phosphate kinase (PIPKII) is an enzyme responsible for the synthesis of phosphatidylinositol-4,5-bisphosphate (PI-4,5-P(2)) from phosphatidylinositol-5-phosphate (PI-5-P). In this study, we demonstrate the presence of PIPKII alpha in bovine photoreceptor rod outer segments (ROS) and the involvement of tyrosine phosphorylation in the regulation of its activity. PIPKII activity in bovine ROS was verified by the preferential conversion of synthetic dipalmitoyl PI-5-P to PI-4,5-P(2), lack of effect of phosphatidic acid, inhibition by heparin, immunoreaction with an anti-PIPKII alpha antibody on Western blots, and immunocytochemical localization in bovine and rat ROS by anti-PIPKII alpha. Immunoprecipitates of bovine ROS with the anti-PIPKII alpha antibody possessed PIPK enzymatic activity and preferentially used PI-5-P as substrate for PI-4,5-P(2) biosynthesis. The activity of PIPKII was greatly increased under conditions favoring tyrosine phosphorylation in ROS, and PIPKII activity was immunoprecipitated with anti-phosphotyrosine (anti-PY) antibodies from tyrosine phosphorylated ROS. Preincubation of ROS with tyrosine kinase inhibitors almost abolished the kinase activity in the anti-PY immunoprecipitates. Immunoblot analysis showed that PIPKII alpha was present in anti-PY immunoprecipitates from phosphorylated ROS but not from nonphosphorylated controls. We conclude that PIPKII alpha is present in ROS and that its activity is regulated by tyrosine phosphorylation.