EMAIL THIS PAGE TO A FRIEND
Cytogenetics and cell genetics
Assignment of SLC17A6 (alias DNPI), the gene encoding brain/pancreatic islet-type Na+-dependent inorganic phosphate cotransporter to human chromosome 11p14.3.
PMID 11306821