EMAIL THIS PAGE TO A FRIEND
Journal of agricultural and food chemistry
Toxico-kinetics, recovery, and metabolism of napropamide in goats following a single high-dose oral administration.
PMID 11308331
Abstract
Toxicokinetic behavior, recovery and metabolism of napropamide (a pre-emergent herbicide) and its effect on Cytochrome P(450) of liver microsomal pellet were studied following a single high-dose oral administration of 2.5 g kg(-1) and continuous (7 days) oral administration of 500 mg kg(-1) in black Bengal goat. Napropamide was detected in blood at 15 min and the maximum quantity was recovered at 3 h after administration. The absorption rate constant (Ka) value was low indicating poor absorption from the gastrointestinal tract. High elimination half-life (t(1/2) beta) and low body clearance (Cl(B)) values coupled with higher transfer of compound from tissue to central compartment (K(21)) suggest that napropamide persisted in the blood for a long time, i.e., after 72 h of oral administration. The recovery percentage of napropamide, including metabolites, from goats varied from 75.94 to 80.08 and excretion of the parent compound through feces varied from 18.86 to 21.59%, indicating that a major portion of the orally administered napropamide was absorbed from the gastrointestinal tract of goat. Napropamide significantly increased the Cytochrome P(450) content of liver microsomal pellet. The recovery of metabolites from feces, urine, and tissues ranged from 4.2--6.2, 40.81--49.42, and 2.7--11.6%, respectively, during a 4--7 day period. The material balance of napropamide (including metabolites) following a single high-dose oral administration at 2.5 g kg(-1) during 4--7 days after dosing was found to be in the range of 75--80%.
Related Materials