EMAIL THIS PAGE TO A FRIEND
Current opinion in plant biology
Metabolism of tyrosine and tryptophan--new genes for old pathways.
PMID 11312134
Abstract
Tyrosine and tryptophan are precursors for the plant defense compounds dhurrin and indole glucosinolates, respectively. In addition, tryptophan is a precursor for the essential phytohormone indole-3-acetic acid. Recent advances in understanding the biosynthesis of these compounds have come from the characterization of enzymes that catalyze the N-hydroxylation of the precursor amino acid to the oxime intermediate. Furthermore, enzymes catalyzing subsequent biosynthetic steps have also been identified.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
07743
Dhurrin, ≥95% (HPLC)
C14H17NO7