EMAIL THIS PAGE TO A FRIEND
Journal of agricultural and food chemistry
NMR and MS analysis of decomposition compounds produced from N-acetyl-L-glutamine at low pH.
PMID 11312773
Abstract
N-Acetyl-L-glutamine decomposition products glutamine, glutamic acid, pyroglutamic acid, N-acetylglutamic acid, and a novel compound, N-(2,6-dioxo-3-piperidinyl) acetamide, have been identified by NMR and MS techniques. N-Acetylglutamine, a modified amino acid, offers greater chemical stability than glutamine in conditions that are experienced during typical sterilization and shelf storage of liquid nutritionals. However, to support safety and stability studies, potential decomposition products of N-acetyl-L-glutamine needed to be identified. Therefore, atypically harsh conditions were used; an unbuffered (pH <3) 1 mg/mL water solution of N-acetyl-L-glutamine was heated to 100 degrees C for 3 h. One-dimensional proton and proton-decoupled carbon-13 NMR and electrospray LC-MS/MS techniques were employed to identify the molecular structures of the generated N-acetyl-L-glutamine decomposition products. Additionally, DEPT and two-dimensional NMR techniques TOCSY, GMQCOSY, GHSQC, and GHMBC were employed to derive the final structure of the acetamide.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A9125
Nα-Acetyl-L-glutamine
C7H12N2O4