EMAIL THIS PAGE TO A FRIEND
The Veterinary record
Inclusion of dimetridazole in gamebird feeds.
PMID 11321219
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D4025
Dimetridazole
C5H7N3O2
31707
Dimetridazole, VETRANAL, analytical standard
C5H7N3O2