EMAIL THIS PAGE TO A FRIEND
Journal of clinical microbiology
Rapid pulsed-field gel electrophoresis protocol for subtyping of Campylobacter jejuni.
PMID 11326009
Abstract
We developed a rapid pulsed-field gel electrophoresis (PFGE) protocol for subtyping Campylobacter isolates based on the standardized protocols used by PulseNet laboratories for the subtyping of other food-borne bacterial pathogens. Various combinations of buffers, reagents, reaction conditions (e.g., cell suspension concentration, lysis time, lysis temperature, and restriction enzyme concentration), and electrophoretic parameters were evaluated in an effort to devise a protocol that is simple, rapid, and robust. PFGE analysis of Campylobacter isolates can be completed in 24 to 30 h using this protocol, whereas the most widely used current protocols require 3 to 4 days to complete. Comparison of PFGE patterns obtained in six laboratories showed that subtyping results obtained using this protocol are highly reproducible.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SMAI-RO Sma I, from Serratia marcescens Sb