EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
1,2,5-Thiadiazole derivatives are potent and selective ligands at human 5-HT1A receptors.
PMID 11327592
Abstract
Amino acid derivatives of 1,2,5-thiadiazol-3-yl-piperazine related to (+)-WAY-100135 and WAY-100635 are potent 5-HT1A receptor agonists and antagonists, which have selective affinity for 5-HT1A receptors versus alpha1 and dopamine (D2, D3, and D4) receptors.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
258903
3,4-Dichloro-1,2,5-thiadiazole, 97%
C2Cl2N2S