EMAIL THIS PAGE TO A FRIEND
FASEB journal : official publication of the Federation of American Societies for Experimental Biology
Dystonia and cerebellar atrophy in Cacna1a null mice lacking P/Q calcium channel activity.
PMID 11344116
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C1426
Cesium methanesulfonate, used in patch clamp techniques
CH3CsO3S