EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
Reductive cleavage of heteroaryl c-halogen bonds by iodotrimethylsilane. Facile and regioselective dechlorination of antibiotic pyrrolnitrin.
PMID 11348155
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
195529
Iodotrimethylsilane, 97%
C3H9ISi