EMAIL THIS PAGE TO A FRIEND
Organic letters
Asymmetric strecker synthesis of alpha-amino acids via a crystallization-induced asymmetric transformation using (R)-phenylglycine amide as chiral auxiliary.
PMID 11348174
Abstract
[reaction: see text]. Diastereoselective Strecker reactions based on (R)-phenylglycine amide as chiral auxiliary are reported. The Strecker reaction is accompanied by an in situ crystallization-induced asymmetric transformation, whereby one diastereomer selectively precipitates and can be isolated in 76-93% yield and dr > 99/1. The diastereomerically pure alpha-amino nitrile obtained from pivaldehyde (R1 = t-Bu, R2 = H) was converted in three steps to (S)-tert-leucine in 73% yield and >98% ee.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
399000
2,2,2-Trifluoroethyl butyrate, 98%
C6H9F3O2
330469
N-Phenylglycine, 97%
C8H9NO2