EMAIL THIS PAGE TO A FRIEND
Acta crystallographica. Section C, Crystal structure communications
L-Phenylalanyl-L-alanine dihydrate.
PMID 11353256
Abstract
A new type of molecular arrangement for dipeptides is observed in the crystal structure of L-phenylalanyl-L-alanine dihydrate, C12H16N2O3-2H2O. Two L-Phe and two L-Ala side chains aggregate into large hydrophobic columns within a three-dimensional hydrogen-bond network.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P3251
Phe-Ala
C12H16N2O3