EMAIL THIS PAGE TO A FRIEND
Electrophoresis
Medium-throughput pKa screening pharmaceuticals by pressure-assisted capillary electrophoresis.
PMID 11358135
Abstract
A fast screening method for the determination of the dissociation constants (pKa) of acidic, basic, and multivalent compounds was developed by using pressure-assisted capillary electrophoresis (PACE). External air pressure was applied to shorten the analysis time. The separation efficiency decreases as air pressure increases. However, it was found that air pressure does not affect the measurement of electrophoretic mobility and pKa significantly when it is less than 2 psi. The method was evaluated in terms of accuracy, precision, and ruggedness by using a set of 48 compounds with literature pKa values ranging from 2 to 10. The difference between the measured pKa values and literature values is less than 0.2 units. The throughput is approximately 20 compounds per day with a 12-point measurement ranging from pH 2.5 to 11. It was demonstrated that this method is applicable for pKa screening of pharmaceuticals with diverse chemical structures.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
W336807
4-Methyl-3-penten-2-one, 90%
C6H10O
281832
Mesityl oxide, technical grade, 90%
C6H10O