EMAIL THIS PAGE TO A FRIEND
The Journal of biological chemistry
X-linked inhibitor of apoptosis protein (XIAP) inhibits caspase-3 and -7 in distinct modes.
PMID 11359776
Abstract
The inhibitor of apoptosis proteins (IAP) regulates cell death by inhibiting caspases. The region of X-linked (X) IAP containing the second baculovirus IAP repeat domain (BIR2) is sufficient for inhibiting caspase-3 and -7. In this study, we found that the modes of inhibition of these two caspases were different: caspase-3 is inhibited in a competitive manner whereas caspase-7 inhibition occurs through a mixed competitive and noncompetitive mechanism. Binding assays revealed that the inhibition of caspase-3 by XIAP was totally dependent on the interaction between the active site of caspase-3 and the linker region between the BIR1 and BIR2 domains of XIAP. In contrast, the active site and the NH(2)-terminal region of caspase-7 bound to the linker region and the BIR2, respectively. Moreover the BIR2 with a mutated linker region, which inhibited caspase-3 very weakly, still bound to and inhibited caspase-7. Furthermore, a chimeric caspase-7/3 comprising the NH(2)-terminal portion of caspase-7 and COOH-terminal portion of caspase-3 was inhibited by XIAP by a mixed competitive and noncompetitive mechanism. Our results suggest that the linker region between BIR1 and BIR2 domains is responsible for active site-directed, competitive inhibition of both caspase-3 and -7, whereas the BIR2 itself is involved in noncompetitive inhibition of caspase-7.