EMAIL THIS PAGE TO A FRIEND
Archives of biochemistry and biophysics
FADD null mouse embryonic fibroblasts undergo apoptosis after photosensitization with the silicon phthalocyanine Pc 4.
PMID 11361017
Abstract
Oxidative stress, such as photodynamic therapy with the silicon phthalocyanine Pc 4 (Pc 4-PDT), can induce apoptosis and tumor necrosis factor alpha (TNF) production. TNF receptors, as well as other death receptors, have been implicated in stress-induced apoptosis. To assess directly the role of FADD, a death receptor-associated protein, in induction of apoptosis post-Pc 4-PDT, embryonic fibroblasts from FADD knock out (k/o) and wild-type (wt) mice were used. Pc 4-PDT induced casp-3 activation and apoptosis in both cell types. In the presence of zVAD, a pancaspase inhibitor, Pc 4-PDT-induced apoptosis was abrogated in both cell lines. Fumonisin B1 (FB), an inhibitor of ceramide synthase, had no effect on apoptosis after Pc 4-PDT in either cell line. Similar to Pc 4-PDT, exogenous C6-ceramide bypassed FADD deficiency and induced zVAD-sensitive apoptosis. In contrast to Pc 4 photosensitization, TNF did not induce either apoptosis or ceramide accumulation in FADD k/o cells. In the absence of FADD deficiency, TNF-induced apoptosis was zVAD-sensitive and FB-insensitive. Induced ceramide levels remained elevated after cotreatment with TNF and zVAD in FADD wt cells. Taken together, these data provide genetic evidence for a lack of FADD requirement in Pc 4-PDT- or C6-ceramide-induced apoptosis. FB-sensitive ceramide production accompanies, but does not suffice, for apoptosis after Pc 4 photosensitization or TNF.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
389935
Silicon 2,3-naphthalocyanine bis(trihexylsilyloxide), Dye content 95 %
C84H102N8O2Si3